Skip to Content
Merck
All Photos(1)

Key Documents

SML3686

Sigma-Aldrich

MYF-03-176

≥98% (HPLC)

Synonym(s):

2-Fluoro-1-[(3R,4R)-3-(2-pyrimidinylamino)-4-[[4-(trifluoromethyl)phenyl]methoxy]-1-pyrrolidinyl]-2-propen-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€173.00
25 MG
€695.00

€173.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€173.00
25 MG
€695.00

About This Item

Empirical Formula (Hill Notation):
C19H18F4N4O2
CAS Number:
Molecular Weight:
410.37
UNSPSC Code:
12352200
NACRES:
NA.21

€173.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MYF-03-176 is an orally active, potent, irreversible pan-TEAD inhibitor that disrupts YAP-TEAD association by targeting TEAD palmitate pocket for covalent binding. MYF-03-176 suppresses NCI-H226 cellular TEAD transcriptional activity (IC50 = 11 nM by TEAD reporter assay), blocking TEAD-mediated target genes expression (CTGF, CYR61, ANKRD1) and suppression (BMF). MYF-03-176 effectively inhibits Hippo signaling-driven human lung mesothelioma NCI-H226 proliferaton in cultures (IC50 = 9 nM in 5 days) and in a xenograft model in mice in vivo (35 or 75 mg/kg b.i.d. p.o. for 4 wks).
Orally active, potent, irreversible pan-TEAD inhibitor that disrupts YAP-TEAD association and inhibits Hippo signaling-driven cancer proliferation in vitro and in vivo.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalent disruptor of YAP-TEAD association suppresses defective Hippo signaling
eLife, 11, e78810-e78810 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service