Skip to Content
Merck
All Photos(1)

Key Documents

SML3677

Sigma-Aldrich

Cryosim-3

≥98% (HPLC)

Synonym(s):

1-Di(isopropyl)phosphinoyl-nonane, 1-Diisopropylphosphorylnonane, Bis(1-methylethyl)nonylphosphine oxide, DIPA 1-9

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€144.00
25 MG
€579.00

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€144.00
25 MG
€579.00

About This Item

Empirical Formula (Hill Notation):
C15H33OP
CAS Number:
Molecular Weight:
260.40
UNSPSC Code:
12352200
NACRES:
NA.25

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

2-8°C

SMILES string

O=P(C(C)C)(C(C)C)CCCCCCCCC

InChI key

DHOTYMJGZIUWRT-UHFFFAOYSA-N

Biochem/physiol Actions

Cryosim-3 is a water soluble, selective and potent agonist of TRPM8 (transient receptor potential melastatin 8) receptor. Cryosim-3 increases tear secretion in an animal model of dry eye disease. Also it relieves ocular discomfort in subjects diagnosed with DED without eye irritation.
Water soluble, selective and potent agonist of TRPM8 receptor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Yin et al.
Science (New York, N.Y.), 378(6616), eadd1268-eadd1268 (2022-10-14)
The transient receptor potential melastatin 8 (TRPM8) channel is the primary molecular transducer responsible for the cool sensation elicited by menthol and cold in mammals. TRPM8 activation is controlled by cooling compounds together with the membrane lipid phosphatidylinositol 4,5-bisphosphate (PIP2).
Jee Myung Yang et al.
BMC ophthalmology, 17(1), 101-101 (2017-06-28)
Physical cooling of the eye surface relieves ocular discomfort, but translating this event to drug treatment of dry eye discomfort not been studied. Here, we synthesized a water-soluble TRPM8 receptor agonist called cryosim-3 (C3, 1-diisopropylphosphorylnonane) which selectively activates TRPM8 (linked

Questions

  1. Could you please tell me the molar concentration of this compound? And the volume of it is too little to absorb.

    1 answer
    1. This product is a neat or pure oil. The density of this material is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service