Skip to Content
Merck
All Photos(1)

Key Documents

SML3581

Sigma-Aldrich

NVP-ACQ090

≥98% (HPLC)

Synonym(s):

1-[[(4S,4aS,8aR)-Decahydro-2-[(2S)-3-(6-methoxy-3-pyridinyl)-2-methylpropyl]-4-isoquinolinyl]carbonyl]-4-(3,4-difluorophenyl)-piperazine, ACQ 090, ACQ-090, ACQ090, [(4S,4aS,8aR)-Decahydro-2-[(2S)-3-(6-methoxy-3-pyridinyl)-2-methylpropyl]-4-isoquinolinyl][4-(3,4-difluorophenyl)-1-piperazinyl]methanone, [4-(3,4-Difluoro-phenyl)-piperazine-1-yl]-{(4S,4aS,8aR)-2[(S)-3-(6-methoxy-pyridin-3-yl)-2-methyl-propyl]-decahydroisoquinoline-4-yl}-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€75.80
25 MG
€308.00

€75.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€75.80
25 MG
€308.00

About This Item

Empirical Formula (Hill Notation):
C30H40F2N4O2
CAS Number:
Molecular Weight:
526.66
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

€75.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

NVP-ACQ090 is a non-peptidic, potent and subtype-selective somatostatin (SRIF; somatotropin release-inhibiting factor) receptor (SST3; SSTR3) antagonist (hSST3 pKD = 8.15, hSSTR1/2/4/5 pKD = 5.69/5.34/5.83/5.95). NVP-ACQ090 blocks hSST3 phosphorylation induced by somatostatin-14 (SS-14) in cultures and impaires object recognition in wild-type mice in vivo (50 μg/kg i.p.).
Non-peptidic, potent and subtype-selective somatostatin (SRIF; somatotropin release-inhibiting factor) receptor (SST3; SSTR3) antagonist in vitro and in vivo

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decahydroisoquinoline derivatives as novel non-peptidic, potent and subtype-selective somatostatin sst(3) receptor antagonists
Bioorganic & Medicinal Chemistry Letters, 20(5), 1728-1734 (2010)
Andreas Lehmann et al.
Molecular endocrinology (Baltimore, Md.), 30(6), 645-659 (2016-04-22)
The human somatostatin receptor 3 (sst3) is expressed in about 50% of all neuroendocrine tumors and hence a promising target for multireceptor somatostatin analogs. The sst3 receptor is unique among ssts in that it exhibits a very long intracellular C-terminal
Emily B Einstein et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(12), 4306-4314 (2010-03-26)
Most neurons possess a single, nonmotile cilium that projects out from the cell surface. These microtubule-based organelles are important in brain development and neurogenesis; however, their function in mature neurons is unknown. Cilia express a complement of proteins distinct from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service