Skip to Content
Merck
All Photos(1)

Key Documents

SML3568

Sigma-Aldrich

C26-A6

≥98% (HPLC)

Synonym(s):

5-chloro-2-methoxy-N-(2-methyl-[1,2,4]triazolo[1,5-a]pyridin-8-yl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€95.00
25 MG
€383.00

€95.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€95.00
25 MG
€383.00

About This Item

Empirical Formula (Hill Notation):
C14H13ClN4O3S
CAS Number:
Molecular Weight:
352.80
UNSPSC Code:
12352200
NACRES:
NA.21

€95.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=S(NC1=CC=CN2N=C(C)N=C12)(C3=CC(Cl)=CC=C3OC)=O

InChI key

HLRMNXITZQAUIH-UHFFFAOYSA-N

Biochem/physiol Actions

A26-A6 is a MTDH-SND1 interaction blocker (IC50 = 2.4 μM by cell-free & 12.3 μM by cellular split-luc assay) that competes against metadherin (MTDH; AEG-1; LYRIC) for staphylococcal nuclease domain-containing 1 (SND1) binding, thereby preventing MTDH-SND1 complex from suppressing antitumor T cell responses. A26-A6 inhibits PyMT tumor sphere formation in cultures (IC50 <200 μM) in a MTDH- and SND1-dependent manner and exhibits in vivo efficacy against breast tumor growth and metastasis in mice in vivo (15 mg/kg/d i.v. 5d/wk), including the murine PyMT model, and human breast cancer xenografts (PDX, HCI-001, SCP28).
Metadherin-staphylococcal nuclease domain-containing 1 (MTDH-SND1) interaction inhibitor with anti-triple-negative breast cancer (TNBC) efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minhong Shen et al.
Nature cancer, 3(1), 60-74 (2022-02-06)
Despite increased overall survival rates, curative options for metastatic breast cancer remain limited. We have previously shown that metadherin (MTDH) is frequently overexpressed in poor prognosis breast cancer, where it promotes metastasis and therapy resistance through its interaction with staphylococcal
Minhong Shen et al.
Nature cancer, 3(1), 43-59 (2022-02-06)
Metastatic breast cancer is a leading health burden worldwide. Previous studies have shown that metadherin (MTDH) promotes breast cancer initiation, metastasis and therapy resistance; however, the therapeutic potential of targeting MTDH remains largely unexplored. Here, we used genetically modified mice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service