Skip to Content
Merck
All Photos(1)

Key Documents

SML3566

Sigma-Aldrich

SUN11602

≥95% (HPLC)

Synonym(s):

4-[[4-[[2-(4-Amino-2,3,5,6-tetramethylanilino)acetyl]-methylamino]piperidin-1-yl]methyl]benzamide, 4-[[4-[[2-[(4-Amino-2,3,5,6-tetramethylphenyl)amino]acetyl]methylamino]-1-piperidinyl]methyl]benzamide, SUN 11602, SUN-11602

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O2
CAS Number:
Molecular Weight:
451.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SUN11602 is an orally active basic fibroblast growth factor (bFGF, FGF2) mimetic that protects against glutamate toxicity in primary rat cerebrocortical neuron (0.1-1 μM SUN11602; 150 μM glutamate) and murine hippocampal neuron cultures (0.3-3 μM SUN11602; 100 μM glutamate). When administered in vivo, SUN11602 exhibits therapeutic efficacy in a rat hippocampal-lesion model (1-10 mg/kg p.o. 24h post Aβ1-40 injection) and a murine MPTP-induced nigrostriatal degeneration model (2.5-5 mg/kg/day p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norihito Murayama et al.
Brain research, 1594, 71-81 (2014-12-03)
Basic fibroblast growth factor (FGF-2/bFGF) possesses neuroprotective activity and promotes cell proliferation. In this study, the novel synthetic compound 4-({4-[[(4-amino-2,3,5,6-tetramethylanilino)acetyl](methyl)amino]-1-piperidinyl}methyl)benzamide (SUN11602) exhibited neuroprotective activities similar to those of FGF-2 without promoting cell proliferation. In primary cultures of hippocampal neurons, stimulation
Ryoko Ogino et al.
Brain research, 1585, 159-166 (2014-08-19)
Basic fibroblast growth factor (bFGF/FGF-2) is known to possess neuroprotective and neurite outgrowth activity properties. In this study, the effects of a novel synthetic compound that mimics the neuroprotective properties of bFGF - SUN11602 - were examined in vitro and
Baojiang Chen et al.
Bioengineered, 13(1), 1530-1540 (2022-01-11)
Psoriasis is a common chronic immune-mediated disease that often has a serious negative impact on the physical and mental health of patients. Dihydroartemisinin (DHA) is a drug with anti-fibrotic and anti-inflammatory effects that may be involved in the autoimmune regulation
Norihito Murayama et al.
ACS chemical neuroscience, 4(2), 266-276 (2013-02-21)
Basic fibroblast growth factor (bFGF) offers some measure of protection against excitotoxic neuronal injuries by upregulating the expression of the calcium-binding protein calbindin-D28k (Calb). The newly synthesized small molecule 4-({4-[[(4-amino-2,3,5,6-tetramethylanilino)acetyl](methyl)amino]-1-piperidinyl}methyl)benzamide (SUN11602) mimics the neuroprotective effects of bFGF, and thus, we
SUN11602, a bFGF mimetic, modulated neuroinflammation, apoptosis and calcium-binding proteins in an in vivo model of MPTP-induced nigrostriatal degeneration
Journal of Neuroinflammation, 19(1), 107-107 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service