SML3564
ML382
≥98% (HPLC)
Synonym(s):
VU0485891, 2-(Cyclopropanesulfonamido)-N-(2-ethoxyphenyl)benzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=C(C=CC=C1)NS(C2CC2)(=O)=O)NC3=C(C=CC=C3)OCC
Biochem/physiol Actions
ML382 is a positive allosteric modulator of Mas-related G-protein-coupled receptor X1 (MrgprX1, MrgX1, SNSR4), a G-protein coupled receptor (GPCR) selectively expressed in dorsal root ganglion (DRG) neurons. MrgX1 is believed to be a functional ortholog of mouse MrgC11, which has been shown to be involved in itch and pain. ML382 is selective for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters, with an EC50 value of 190 nm with an Emax value of 148% in the presence of the known ligand BAM8-22.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Discovery and characterization of 2-(cyclopropanesulfonamido)-N-(2-ethoxyphenyl)benzamide, ML382: a potent and selective positive allosteric modulator of MrgX1
ChemMedChem, 10, 57-61 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service