Skip to Content
Merck
All Photos(1)

Key Documents

SML3544

Sigma-Aldrich

Ginsenoside Ro

≥94% (HPLC)

Synonym(s):

(3β)-28-(β-D-Glucopyranosyloxy)-28-oxoolean-12-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranosiduronic acid, Chikusetsusaponin 5, Polysciasaponin P3

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€125.00
25 MG
€502.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€125.00
25 MG
€502.00

About This Item

Empirical Formula (Hill Notation):
C48H76O19
CAS Number:
Molecular Weight:
957.11
UNSPSC Code:
12352200
NACRES:
NA.77

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥94% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O)C(=O)O)O[C@@H]3C([C@H]4[C@@]([C@@H]5[C@]([C@@]6(CC[C@@]8([C@@H](CC(CC8)(C)C)C6=CC5)C(=O)O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7O)O)O)CO)C)(CC4)C)(CC3)C)(C)C

InChI

1S/C48H76O19/c1-43(2)14-16-48(42(61)67-40-35(58)31(54)29(52)24(20-50)63-40)17-15-46(6)21(22(48)18-43)8-9-26-45(5)12-11-27(44(3,4)25(45)10-13-47(26,46)7)64-41-37(33(56)32(55)36(65-41)38(59)60)66-39-34(57)30(53)28(51)23(19-49)62-39/h8,22-37,39-41,49-58H,9-20H2,1-7H3,(H,59,60)/t22-,23+,24+,25-,26+,27-,28+,29+,30-,31-,32-,33-,34+,35+,36-,37+,39-,40-,41+,45-,46+,47+,48-/m0/s1

InChI key

NFZYDZXHKFHPGA-QQHDHSITSA-N

Related Categories

Biochem/physiol Actions

Ginsenoside Ro is a key constituent of ginseng that exhibits varies health beneficial activities including antitumor, anti-inflammatory, and antioxidant. Ginsenoside Ro ameliorates obesity and insulin resistance in high fat diet-induced obese (DIO) mice without affecting food intake through activation of G protein-coupled bile acid receptor 5 (TGR5).
TGR5 activator; key constituent of ginseng that exhibits varies health beneficial activities including antitumor, anti-inflammatory, and antioxidant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginsenoside Ro Ameliorates High-Fat Diet-Induced Obesity and Insulin Resistance in Mice via Activation of the G Protein-Coupled Bile Acid Receptor 5 Pathway
Journal of Pharmacology and Experimental Therapeutics, 377, 441-451 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service