Skip to Content
Merck
All Photos(1)

Key Documents

SML3530

Sigma-Aldrich

DMA-135

≥98% (HPLC)

Synonym(s):

3-​amino-​N-​(aminoiminomethyl)​-​5-​(dimethylamino)​-​6-​(2-​phenylethynyl)​-2-​pyrazinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€69.80
25 MG
€282.00

€69.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€69.80
25 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C16H17N7O · xHCl
CAS Number:
Molecular Weight:
323.35 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€69.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=C(N=C(C(N)=N1)C(NC(N)=N)=O)C#CC2=CC=CC=C2)C.[xHCl]

Biochem/physiol Actions

Cell penetrant and potent inhibitor of human enterovirus 71 (EV71) translation and replication that targets the IRES domain of EV71
DMA-135 is a cell penetrant and potent inhibitor of human enterovirus 71 (EV71) translation and replication that targets the IRES domain of EV71. DMA-135 binds allosterically to EV71 SLII RNA and stabilizes ternary complex consisting of AUF1-SLII-DMA-135. Also, previous data indicate that DMA-135 exhibit comparable affinity to HIV-1-TAR and HIV-2-TAR.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amiloride as a new RNA-binding scaffold with activity against HIV-1 TAR
MedChemComm, 8(5), 1022-1036 (2017)
IRES-targeting small molecule inhibits enterovirus 71 replication via allosteric stabilization of a ternary complex
Nature Communications, 11(1), 4775-4775 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service