Skip to Content
Merck
All Photos(1)

Key Documents

SML3473

Sigma-Aldrich

144DG11

≥98% (HPLC)

Synonym(s):

1,2-Dihydro-N-(2-methoxyethyl)-6-methyl-N-[(3-methyl-2-thienyl)methyl]-2-oxo-4-pyridinecarboxamide, 2-Hydroxy-N-(2-methoxyethyl)-6-methyl-N-((3-methylthiophen-2-yl)methyl)isonicotinamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.50
25 MG
€225.00

€55.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.50
25 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C16H20N2O3S
CAS Number:
Molecular Weight:
320.41
UNSPSC Code:
12352200
NACRES:
NA.77

€55.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder or solid

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

application(s)

research use

storage temp.

2-8°C

SMILES string

OC1=NC(C)=CC(C(N(CCOC)CC2=C(C)C=CS2)=O)=C1

InChI key

BBEZNLRXWCUHAJ-UHFFFAOYSA-N

Biochem/physiol Actions

144DG11 is a polyglucosan (PG)-reducing agent that targets the lysosomal membrane protein LAMP1 and increases autophagic flux in adult polyglucosan body disease (APBD) primary fibroblasts (50 μM for 24 hrs), resulting in an enhanced glycophagy, respiratory and glycolytic ATP production. 144DG11 prolongs survival and improves motor parameters of Gbe(ys/ys) mice in vivo (250 mg/150 μL/kg 2x per wk via i.v. for 1 month, then s.c.), a murine APBD model, by reducing PG and glycogen levels in brain, liver, heart, and peripheral nerve with good pharmacokinetic properties and no adverse effects to the animals.
Polyglucosan (PG)-reducing agent that targets the lysosomal membrane protein LAMP1 in cultures and in vivo.

Disclaimer

For research use only

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service