Skip to Content
Merck
All Photos(1)

Key Documents

SML3458

Sigma-Aldrich

KL-11743

≥98% (HPLC)

Synonym(s):

2-(3-(6-Ethoxy-4-((4-(1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazol-4-yl)phenyl)amino)quinazolin-2-yl)phenoxy)-N-isopropylacetamide, 2-[3-[6-Ethoxy-4-[[4-(1H-pyrazol-4-yl)phenyl]amino]-2-quinazolinyl]phenoxy]-N-(1-methylethyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€76.40
25 MG
€310.00

€76.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€76.40
25 MG
€310.00

About This Item

Empirical Formula (Hill Notation):
C30H30N6O3
CAS Number:
Molecular Weight:
522.60
UNSPSC Code:
12352200
NACRES:
NA.77

€76.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

KL-11743 is an orally available, potent inhibitor of class I glucose transporters that blocks glucose uptake in vitro and in vivo. It induces aspartate accumulation and increases mitochondrial metabolism. KL-11743 specifically inhibits the growth of tumors containing truncated the tricarboxylic acid (TCA) cycle due to mutations inactivating succinate dehydrogenase. Inhibition of glucose uptake by KL-11743 may synergize with therapies that induce oxidative stress (synthetic lethality).
orally available, potent inhibitor of class I glucose transporters that blocks glucose uptake in vitro and in vivo

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service