Skip to Content
Merck
All Photos(1)

Key Documents

SML3439

Sigma-Aldrich

IXA4

98% (HPLC)

Synonym(s):

N-​Methyl-​N-​[2-​(4-​methylphenoxy)​ethyl]​-​4-​[(1-​oxo-​3-​phenoxypropyl)​amino]​-1H-​pyrazole-​1-​acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.90
25 MG
€226.00

€55.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.90
25 MG
€226.00

About This Item

Empirical Formula (Hill Notation):
C24H28N4O4
CAS Number:
Molecular Weight:
436.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€55.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CN1N=CC(NC(CCOC2=CC=CC=C2)=O)=C1)N(C)CCOC3=CC=C(C=C3)C

InChI

1S/C24H28N4O4/c1-19-8-10-22(11-9-19)32-15-13-27(2)24(30)18-28-17-20(16-25-28)26-23(29)12-14-31-21-6-4-3-5-7-21/h3-11,16-17H,12-15,18H2,1-2H3,(H,26,29)

InChI key

ZVSKMVAWWBSNOY-UHFFFAOYSA-N

Biochem/physiol Actions

IXA4 is a nontoxic, potent and highly selective IRE1-XBP1s signaling activator that induce ER proteostasis remodeling. IXA4 reduced Aβ levels and prevents APP-associated mitochondrial dysfunction in CHO7PA2 cells expressing the V717F APP (APPV717F) mutant.
Nontoxic, potent and highly selective IRE1-XBP1s signaling activator that induce ER proteostasis remodeling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service