Skip to Content
Merck
All Photos(1)

Key Documents

SML3366

Sigma-Aldrich

SIS17

≥95% (HPLC)

Synonym(s):

N′-Hexadecylthiophene-2-carbohydrazide, SIS 17, SIS-17

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€77.00
50 MG
€254.00

€77.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€77.00
50 MG
€254.00

About This Item

Empirical Formula (Hill Notation):
C21H38N2OS
CAS Number:
Molecular Weight:
366.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€77.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[s]1c(ccc1)C(=O)NNCCCCCCCCCCCCCCCC

InChI

1S/C21H38N2OS/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18-22-23-21(24)20-17-16-19-25-20/h16-17,19,22H,2-15,18H2,1H3,(H,23,24)

InChI key

HSHXDCVZWHOWCS-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective HDAC11 inhibitor that effectively upregulates cellular serine hydroxymethyl transferase 2 (SHMT2), fatty acylation level.
SIS17 is a potent and selective HDAC11 inhibitor (IC50 = 830 nM; IC50 >100 μM against HDAC1/4/8 or SIRT1/2/3/6) that effectively upregulates HDAC11 substrate, serine hydroxymethyl transferase 2 (SHMT2), fatty acylation level in MCF7 cells (12.5 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service