Skip to Content
Merck
All Photos(1)

Key Documents

SML3337

Sigma-Aldrich

CBR-470-1, racemic

≥98% (HPLC)

Synonym(s):

(± )-3-((4-chlorophenyl)sulfonyl)-4-(isobutylamino)tetrahydrothiophene 1,1-dioxide, 3-Thiophenamine, 4-[(4-chlorophenyl)sulfonyl]tetrahydro-N-(2-methylpropyl)-, 1,1-dioxide, (3R,4S)-rel- (ACI)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€123.00
25 MG
€395.00

€123.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€123.00
25 MG
€395.00

About This Item

Empirical Formula (Hill Notation):
C14H20ClNO4S2
CAS Number:
Molecular Weight:
365.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€123.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2%, clear

storage temp.

2-8°C

Biochem/physiol Actions

CBR-470-1 is an Inhibitor of the glycolytic enzyme PGK1 that induces Nrf2 activity. PGK1 inhibition results in acculumation of methylglyoxalyl (MGO), a toxic, reactive metabolite removed by other enzymes. The accumulated MGO then irreversibly reacts with Keap1 forming inactive Keap1 dimers, resulting in accumulation of Nrf2.
Inhibitor of the glycolytic enzyme PGK1 inducing Nrf2 activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeyun Jo et al.
Bioorganic chemistry, 108, 104614-104614 (2021-01-29)
The transcription factor NRF2 controls resistance to oxidative insult and is thus a key therapeutic target for treating a number of disease states associated with oxidative stress and aging. We previously reported CBR-470-1, a bis-sulfone which activates NRF2 by increasing
Jinyu Zheng et al.
Aging, 12(13), 13388-13399 (2020-07-11)
The neurotoxin MPP+ (1-methyl-4-phenylpyridinium ion) disrupts mitochondrial function leading to oxidative stress and neuronal death. Here we examine whether activation of the Keap1-Nrf2 cascade can protect SH-SY5Y neuroblastoma cells from MPP+-induced cytotoxicity. Treatment of SH-SY5Y cells with CBR-470-1, an inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service