Skip to Content
Merck
All Photos(1)

Key Documents

SML3306

Sigma-Aldrich

BAY-390

≥98% (HPLC)

Synonym(s):

(1R,2R)-2-(4-Fluorophenylamino)-1-(trifluoromethyl)cyclohexanol, BAY 390, BAY390

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€105.00
25 MG
€424.00

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€105.00
25 MG
€424.00

About This Item

Empirical Formula (Hill Notation):
C13H15F4NO
CAS Number:
Molecular Weight:
277.26
UNSPSC Code:
12352200
NACRES:
NA.77

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

yellow to orange

storage temp.

2-8°C

SMILES string

FC1=CC=C(C=C1)N[C@H]2[C@@](C(F)(F)F)(CCCC2)O

InChI key

IESAJAZKMLPVIB-VXGBXAGGSA-N

Biochem/physiol Actions

BAY-390 is an orally active, potent and selective TRPA1 inhibitor (cellular Ca++ influx IC50 in nM by FLIPR = 16/hu, 19/monkey, 63/rat, 68/guinea pig, 73/mu, 81/dog; IC50 = 5.6 µM/hTRPC5, >25 µM/human TRPV1/4, TRPC3/6, KCNK9 (TASK-3), CACNA1H (Cav3.2)) that shows in vivo efficacy in animal models of inflammatory pain (30 or 90 mg/kg mice, p.o.) and neurogenic pain (30 or 90 mg/kg rats, b.i.d. p.o.).
Oally active, potent and selective TRPA1 inhibitor with in vivo efficacy in inflammatory and neurogenic pain models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service