Skip to Content
Merck
All Photos(1)

Key Documents

SML3301

Sigma-Aldrich

cisVZ185

≥98% (HPLC)

Synonym(s):

(2S,4S)-N-(2-(5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyloxy)-4-(4-methylthiazol-5-yl)benzyl)-1-((S)-2-(1-fluorocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide, Cis-VZ 185, Cis-VZ-185, Cis-VZ185, VZ 185 inactive control, VZ 185 negative control, VZ-185 inactive control, VZ185 inactive control, VZ185 negative control

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€222.00
25 MG
€892.00

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€222.00
25 MG
€892.00

About This Item

Empirical Formula (Hill Notation):
C53H67FN8O8S
CAS Number:
Molecular Weight:
995.21
UNSPSC Code:
51111800
NACRES:
NA.32

€222.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NCC1=C(OCCCCCN2CCN(CC3=C(OC)C=C(C(C4=C5C=NC=C4)=CN(C)C5=O)C=C3OC)CC2)C=C(C=C1)C6=C(C)N=CS6)[C@H]7N(C([C@@H](NC(C8(F)CC8)=O)C(C)(C)C)=O)C[C@@H](O)C7

InChI key

ZAGCLFXBHOXXEN-ZLGHNAGESA-N

Biochem/physiol Actions

Inactive cis-isomer of and the negative control for the potent and selective PROTAC BRD7/9 degrader VZ185.
cisVZ185 is the inactive cis-isomer of and the recommended negative control for the potent and selective dual BRD7 and BRD9 degrader VZ185.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service