Skip to Content
Merck
All Photos(1)

Key Documents

SML3225

Sigma-Aldrich

trans-Clovamide

≥98% (HPLC)

Synonym(s):

(S,E)-3-(3,4-dihydroxyphenyl)-2-(3-(3,4-dihydroxyphenyl)acrylamido)propanoic acid, N-[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]-3-hydroxy-L-tyrosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€62.30
25 MG
€272.00

€62.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€62.30
25 MG
€272.00

About This Item

Empirical Formula (Hill Notation):
C18H17NO7
CAS Number:
Molecular Weight:
359.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€62.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C([C@H](CC1=CC(O)=C(C=C1)O)NC(/C=C/C2=CC(O)=C(C=C2)O)=O)O

InChI

1S/C18H17NO7/c20-13-4-1-10(8-15(13)22)3-6-17(24)19-12(18(25)26)7-11-2-5-14(21)16(23)9-11/h1-6,8-9,12,20-23H,7H2,(H,19,24)(H,25,26)/b6-3+/t12-/m0/s1

InChI key

GPZFXSWMDFBRGS-UXONFWTHSA-N

Biochem/physiol Actions

Clovamide is a phenolic compound isolated from cocoa beans that exhibits antioxidant and neuroprotective effects. Clovamide is a potent and selective inhibitor of Aβ42 aggregation.
inhibitor of Ab42 aggregation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service