Skip to Content
Merck
All Photos(1)

Key Documents

SML3217

Sigma-Aldrich

PF-06843195

≥98% (HPLC), storage temp.:2-8°C

Synonym(s):

2,2-Difluoroethyl (3S)-3-[[2′-amino-5-fluoro-2-(4-morpholinyl)[4,5′-bipyrimidin]-6-yl]amino]-3-(hydroxymethyl)-1-pyrrolidinecarboxylate, PF 06843195, PF 06862309 active drug, PF-06862309 active drug, PF06843195

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€62.30
25 MG
€252.00

€62.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€62.30
25 MG
€252.00

About This Item

Empirical Formula (Hill Notation):
C20H25F3N8O4
CAS Number:
Molecular Weight:
498.46
UNSPSC Code:
12352107
NACRES:
NA.77

€62.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC[C@]1(CN(CC1)C(OCC(F)F)=O)NC2=NC(N3CCOCC3)=NC(C4=CN=C(N)N=C4)=C2F

InChI key

RTOREZYNLPQUKM-FQEVSTJZSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PZ0313PZ0326SML3314
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige

Biochem/physiol Actions

PF-06843195 is a potent and PI3K-alpha subtype-selective phosphoinositide 3-kinase (PI3K) inhibitor (rat PI3K Ki = 2 nM/β, <18 nM/α, <280 nM/δ) that inhibits cellular PI3Kα activity at a higher potency over that of PI3Kδ/β and mTOR (AKT pT308 IC50 = 18/160/360 nM in rat PI3Kα/δ/β Rat1 lines; p70S6K pT389 IC50 = 1.5 μM in Rat1 myristoylated PI3K line). PF-06843195 is more potent against cellular AKT pT308 (MCF7/T47D IC50 = 7.8/8.7 nM) and proliferation (MCF7/T47D = 62/32 nM) than alpelisib (BYL719) in cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service