Skip to Content
Merck
All Photos(1)

Key Documents

SML2803

Sigma-Aldrich

Tebanicline hydrochloride

≥98% (HPLC)

Synonym(s):

(R)-5-(2-Azetidinylmethoxy)-2-chloropyridine hydrochloride, (R)-5-(Azetidin-2-ylmethoxy)-2-chloropyridine hydrochloride, 5-((2R)-Azetidin-2-ylmethoxy)-2-chloropyridine hydrochloride, ABT-594 hydrochloride, Ebanicline hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00
25 MG
€348.00

€108.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€108.00
25 MG
€348.00

About This Item

Empirical Formula (Hill Notation):
C9H11ClN2O · xHCl
CAS Number:
Molecular Weight:
198.65 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1ncc(cc1)OC[C@@H]2NCC2

InChI

1S/C9H11ClN2O/c10-9-2-1-8(5-12-9)13-6-7-3-4-11-7/h1-2,5,7,11H,3-4,6H2/t7-/m1/s1

InChI key

MKTAGSRKQIGEBH-SSDOTTSWSA-N

Biochem/physiol Actions

Partial agonist at neural nicotinic acetylcholine receptors, binding to both the α3β4 and the α4β2 subtypes
Tebanicline (ABT-594) is a partial agonist at neural nicotinic acetylcholine receptors, binding to both the α3β4 and the α4β2 subtypes. It is a less toxic analog of epibatidine with potent antinociceptive effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyong Ding et al.
Psychopharmacology, 235(4), 1093-1105 (2018-01-15)
A novel rodent continuous performance test (CPT) was developed as one of the goals of the NEWMEDS (Novel Methods leading to New Medications in Depression and Schizophrenia) consortium to improve its translatability to the CPT test used in human subjects. The
Takafumi Hayashi et al.
Neuroscience letters, 638, 46-50 (2016-12-13)
Tebanicline (ABT-594), an analogue of epibatidine, exhibits potent antinociceptive effects and high affinity for the nicotinic acetylcholine receptor in the central nervous system. We assessed whether tebanicline exerts an effect on various noxious stimuli and mediates the nicotine receptor or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service