Skip to Content
Merck
All Photos(1)

Key Documents

SML2771

Sigma-Aldrich

Amonafide

≥98% (HPLC)

Synonym(s):

5-Amino-2-[2-(dimethylamino)ethyl]-1H-benz[de]isoquinoline-1,3(2H)-dione, AS1413, FA 142, NSC 308847, Nafidimide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.90
25 MG
€290.00

€85.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€85.90
25 MG
€290.00

About This Item

Empirical Formula (Hill Notation):
C16H17N3O2
CAS Number:
Molecular Weight:
283.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€85.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]1([c](c2c3c([c]1=O)cccc3cc(c2)N)=O)CCN(C)C

InChI

1S/C16H17N3O2/c1-18(2)6-7-19-15(20)12-5-3-4-10-8-11(17)9-13(14(10)12)16(19)21/h3-5,8-9H,6-7,17H2,1-2H3

InChI key

UPALIKSFLSVKIS-UHFFFAOYSA-N

Biochem/physiol Actions

Amonafide, a naphthalimides DNA intercalator, is a potent topoisomerase II inhibitor that inhibits tumor proliferation and progression. It appears that Amonafide inhibits melanoma progression through inhibition of the AKT/mTOR signaling pathway.
DNA intercalator and potent topoisomerase II inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of DNA-intercalative naphthalimide-benzothiazole/cinnamide derivatives: cytotoxicity evaluation and topoisomerase-IIa inhibition
Sankara Rao N, Nagesh N, Lakshma Nayak V, Sunkari S, Tokala R, Kiranmai G, Regur P, et al
MedChemComm, 10, 72-79 (2018)
Ciara L Freeman et al.
Expert review of hematology, 5(1), 17-26 (2012-01-26)
Development of the novel topoisomerase II inhibitor, amonafide, began almost 40 years ago. The drug was selected for further investigation owing to evidence of marked antineoplastic efficacy in preclinical models of cancer. When its usefulness in the treatment of various
Yingyi Ye et al.
Cancer management and research, 11, 2339-2348 (2019-04-10)
Human melanoma is a malignant tumor originated from melanocytes with high invasion, metastasis, and poor prognosis. In this study, the effects of naphthalimides UNBS5162 and amonafide on the properties of proliferation and apoptosis in human melanoma cells were confirmed. Cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service