Skip to Content
Merck
All Photos(1)

Key Documents

SML2683

Sigma-Aldrich

dBET6

≥98% (HPLC)

Synonym(s):

(6S)-4-(4-Chlorophenyl)-N-[8-[[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetyl]amino]octyl]-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€44.00
5 MG
€179.00

€44.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€44.00
5 MG
€179.00

About This Item

Empirical Formula (Hill Notation):
C42H45ClN8O7S
CAS Number:
Molecular Weight:
841.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€44.00


Please contact Customer Service for Availability

Request a Bulk Order

ligand

thalidomide

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c2c(c(c1C)C)C(=N[C@H](c7[n]2c(nn7)C)CC(=O)NCCCCCCCCNC(=O)COc4c5c(ccc4)C(=O)N(C5=O)C6CCC(=O)NC6=O)c3ccc(cc3)Cl

InChI

1S/C42H45ClN8O7S/c1-23-24(2)59-42-35(23)37(26-13-15-27(43)16-14-26)46-29(38-49-48-25(3)50(38)42)21-33(53)44-19-8-6-4-5-7-9-20-45-34(54)22-58-31-12-10-11-28-36(31)41(57)51(40(28)56)30-17-18-32(52)47-39(30)55/h10-16,29-30H,4-9,17-22H2,1-3H3,(H,44,53)(H,45,5

InChI key

JGQPZPLJOBHHBK-UFXYQILXSA-N

Biochem/physiol Actions

dBET6 is a highly cell penetrant, potent and selective BET (bromodomain and extra-terminal domain) degrader that induces rapid (<4hrs) degradation of BRD2, BRD3 and BRD4 and complete down-regulation of c-MYC protein and induction of apoptosis. dBET6 exhibits potent anti-cancer activities.
highly cell penetrant, potent and selective BETdegrader that induces rapid degradation of BRD2, BRD3 and BRD4

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(22), E5086-E5095 (2018-05-17)
Competitive BET bromodomain inhibitors (BBIs) targeting BET proteins (BRD2, BRD3, BRD4, and BRDT) show promising preclinical activities against brain cancers. However, the BET protein-dependent glioblastoma (GBM)-promoting transcriptional network remains elusive. Here, with mechanistic exploration of a next-generation chemical degrader of
Georg E Winter et al.
Molecular cell, 67(1), 5-18 (2017-07-05)
Processive elongation of RNA Polymerase II from a proximal promoter paused state is a rate-limiting event in human gene control. A small number of regulatory factors influence transcription elongation on a global scale. Prior research using small-molecule BET bromodomain inhibitors
Behnam Nabet et al.
Nature chemical biology, 14(5), 431-441 (2018-03-28)
Dissection of complex biological systems requires target-specific control of the function or abundance of proteins. Genetic perturbations are limited by off-target effects, multicomponent complexity, and irreversibility. Most limiting is the requisite delay between modulation to experimental measurement. To enable the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service