Skip to Content
Merck
All Photos(1)

Key Documents

SML2650

Sigma-Aldrich

T3-CLK

≥98% (HPLC)

Synonym(s):

4-(2-Methyl-1-(4-methylpiperazin-1-yl)-1-oxopropan-2-yl)-N-(6-(pyridin-4-yl)imidazo[1,2-a]pyridin-2-yl)benzamide, 4-[1,1-Dimethyl-2-(4-methyl-1-piperazinyl)-2-oxoethyl]-N-[6-(4-pyridinyl)imidazo[1,2-a]pyridin-2-yl]benzamide, T3

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€104.00
25 MG
€419.00

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€104.00
25 MG
€419.00

About This Item

Empirical Formula (Hill Notation):
C28H30N6O2
CAS Number:
Molecular Weight:
482.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€104.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

[n]21c(nc(c2)NC(=O)c4ccc(cc4)C(C)(C)C(=O)N5CCN(CC5)C)ccc(c1)c3ccncc3

InChI

1S/C28H30N6O2/c1-28(2,27(36)33-16-14-32(3)15-17-33)23-7-4-21(5-8-23)26(35)31-24-19-34-18-22(6-9-25(34)30-24)20-10-12-29-13-11-20/h4-13,18-19H,14-17H2,1-3H3,(H,31,35)

InChI key

IEFFSHLHNYVSEF-UHFFFAOYSA-N

Biochem/physiol Actions

T3-CLK (T3) is a cell permeable, highly potent and selective cell-base stable inhibitor of CDC-like kinase (CLK) that exhibits dose-dependent alternative splicing effects in HCT116 colorectal cancer cells. T3-CLK increases conjoined gene (CG) transcription.
cell permeable, highly potent and selective cell-base stable inhibitor of CDC-like kinase (CLK)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susanne Müller et al.
eLife, 7 (2018-04-21)
Potent, selective and broadly characterized small molecule modulators of protein function (chemical probes) are powerful research reagents. The pharmaceutical industry has generated many high-quality chemical probes and several of these have been made available to academia. However, probe-associated data and
Tyler Funnell et al.
Nature communications, 8(1), 7-7 (2017-02-25)
CDC-like kinase phosphorylation of serine/arginine-rich proteins is central to RNA splicing reactions. Yet, the genomic network of CDC-like kinase-dependent RNA processing events remains poorly defined. Here, we explore the connectivity of genomic CDC-like kinase splicing functions by applying graduated, short-exposure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service