Skip to Content
Merck
All Photos(1)

Key Documents

SML2640

Sigma-Aldrich

YZ129

≥98% (HPLC)

Synonym(s):

4-(6-Isoquinolinylamino)-1,2-naphthalenedione, 4-(Isoquinolin-6-ylamino)naphthalene-1,2-dione, YZ-129, YZ129

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€75.90
25 MG
€246.00

€75.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€75.90
25 MG
€246.00

About This Item

Empirical Formula (Hill Notation):
C19H12N2O2
CAS Number:
Molecular Weight:
300.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€75.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint brown-red to dark red

solubility

DMSO: 0.5 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c3cc4c(cncc4)cc3)C1=CC(=O)C(=O)c2c1cccc2

InChI key

NYDMVPHEEBVAHT-UHFFFAOYSA-N

Biochem/physiol Actions

Hsp90 inhibitor with anti-cancer efficacy in vitro and in vivo.
YZ129 is an ATP site-targeting Hsp90 inhibitor (IC50 = 29.5 nM against 2.5 nM geldanamycin-FITC for binding human HSP90α) that prevents thapsigargin (1 μM)-induced NFAT nuclear translocation in HeLa cells (IC50 = 820 nM) by blocking HSP90 client calcineurin-mediated NFAT dephosphorylation. YZ129 (5 μM; 24-48 hr) suppresses U87 glioblastoma (GBM) cell migration and proliferation as a result of G2/M arrest and apoptosis induction. Daily intraperitoneal administration is efficacious against U87 xenografts (s.c.; bilateral posterior limbs)-derived tumor growth in mice in vivo.

Other Notes

Product may exist in a Tautomeric Form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenzhen Liu et al.
Cell chemical biology, 26(3), 352-365 (2019-01-15)
Glioblastoma (GBM) is among the most common and malignant types of primary brain tumors in adults, with a dismal prognosis. Although alkylating agents such as temozolomide are widely applied as the first-line treatment for GBM, they often cause chemoresistance and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service