Skip to Content
Merck
All Photos(1)

Key Documents

SML2620

Sigma-Aldrich

Bromonoscapine

≥98% (HPLC)

Synonym(s):

(S)-3-((R)-9-bromo-4-methoxy-6-methyl-5,6,7,8-tetrahydro-[1,3]dioxolo[4,5-g]isoquinolin-5-yl)-6,7-dimethoxyisobenzofuran-1(3H)-one, 9-Bromonoscapine, EM 011

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€71.60
25 MG
€254.00

€71.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€71.60
25 MG
€254.00

About This Item

Empirical Formula (Hill Notation):
C22H22BrNO7
CAS Number:
Molecular Weight:
492.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€71.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -100 to -120°, c = 1 in dichloromethane

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1c2c(c(c3c1CCN([C@H]3[C@H]4OC(=O)c5c4ccc(c5OC)OC)C)OC)OCO2

InChI

1S/C22H22BrNO7/c1-24-8-7-10-13(19(28-4)21-20(15(10)23)29-9-30-21)16(24)17-11-5-6-12(26-2)18(27-3)14(11)22(25)31-17/h5-6,16-17H,7-9H2,1-4H3/t16-,17+/m1/s1

InChI key

PLMUFLAOTAHIIZ-SJORKVTESA-N

Biochem/physiol Actions

Bromonoscapine is a microtubular disrupting agent that exhibits potent cytotoxic activity against breast, lung and other cancers.
microtubular disrupting agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akhilesh Kumar Verma et al.
Bioorganic & medicinal chemistry, 14(19), 6733-6736 (2006-06-21)
Three haloderivatives of noscapine 2-4 were synthesized chemoselectively and their in vitro cytotoxicity was assessed by MTT assay on U-87 human glioblastoma cell lines. At 50 microM concentration after 72 h, 9-chloronoscapine 2, 9-bromonoscapine 3 (EM011), and 9-iodonoscapine 4 killed
Jun Zhou et al.
Molecular pharmacology, 63(4), 799-807 (2003-03-20)
Noscapine, a microtubule-interfering agent, has been shown to arrest mitosis, to induce apoptosis, and to have potent antitumor activity. We report herein that two brominated derivatives of noscapine, 5-bromonoscapine (5-Br-nosc) and reduced 5-bromonoscapine (Rd 5-Br-nosc), have higher tubulin binding activity
Maged Henary et al.
Biochemical pharmacology, 92(2), 192-205 (2014-08-16)
Noscapine, an opium-derived 'kinder-gentler' microtubule-modulating drug is in Phase I/II clinical trials for cancer chemotherapy. However, its limited water solubility encumbers its development into an oral anticancer drug with clinical promise. Here we report the synthesis of 9 third-generation, water-soluble

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service