Skip to Content
Merck
All Photos(1)

Documents

SML2619

Sigma-Aldrich

THZ531

≥98% (HPLC)

Synonym(s):

(2E)-N-[4-[[(3R)-3-[[5-Chloro-4-(1H-indol-3-yl)-2-pyrimidinyl]amino]-1-piperidinyl]carbonyl]phenyl]-4-(dimethylamino)-2-butenamide, (R,E)-N-(4-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)piperidine-1-carbonyl)phenyl)-4-(dimethylamino)but-2-enamide, THZ 531, THZ-5-31-1, THZ-531

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32ClN7O2
CAS Number:
Molecular Weight:
558.07
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(C/C=C/C(NC1=CC=C(C=C1)C(N2CCC[C@H](C2)NC3=NC(C4=CNC5=C4C=CC=C5)=C(C=N3)Cl)=O)=O)C

Biochem/physiol Actions

THZ531 is a potent, CDK12/13-selective cyclin-dependent kinase inhibitor (IC50 = 158 nM, 69 nM, 10.5 μM, respectively, against 0.2 ?M CDK12-CycK, CDK13-CycK, CDK9-CycT1; IC50 = 8.5 μM against 25 nM CDK7-CycH-MAT1) that targets CDK12 Cys1039 and CDK13 Cys1017 for irreversible covalent modification while bound to the ATP-binding site, displaying good selectivity over >200 other kinases. THZ531 selectively reduces Pol II CTD Ser2, but not Ser5/Ser7, phosphorylation and transcription activity in Jurkat cultures (200-500 nM), exhibiting antiproliferation potency (IC50 = 50 nM in 72 h) due to time- and dose-dependent apoptosis induction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service