Skip to Content
Merck
All Photos(1)

Key Documents

SML2576

Sigma-Aldrich

Neramexane hydrochloride

≥95% (HPLC)

Synonym(s):

1,3,3,5,5-Pentamethylcyclohexanamine hydrochloride, 1-Amino-1,3,3,5,5-pentamethylcyclohexane hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€57.20
50 MG
€288.00

€57.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€57.20
50 MG
€288.00

About This Item

Empirical Formula (Hill Notation):
C11H23N · xHCl
CAS Number:
Molecular Weight:
169.31 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€57.20


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

NC1(CC(CC(C1)(C)C)(C)C)C

InChI

1S/C11H23N/c1-9(2)6-10(3,4)8-11(5,12)7-9/h6-8,12H2,1-5H3

InChI key

OGZQTTHDGQBLBT-UHFFFAOYSA-N

Biochem/physiol Actions

NMDA receptor antagonist
Neramexane is a potent NMDA antagonist that exhibits neuroprotective effects.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service