Skip to Content
Merck
All Photos(1)

Key Documents

SML2544

Sigma-Aldrich

NSC49652

≥98% (HPLC)

Synonym(s):

(2E)-1-(2-Hydroxyphenyl)-3-(3-pyridinyl)-2-propen-1-one, (E)-1-(2-Hydroxyphenyl)-3-(pyridin-3-yl)-2-propen-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€87.30
25 MG
€355.00

€87.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€87.30
25 MG
€355.00

About This Item

Empirical Formula (Hill Notation):
C14H11NO2
CAS Number:
Molecular Weight:
225.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€87.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=CC=C1C(/C=C/C2=CN=CC=C2)=O

InChI

1S/C14H11NO2/c16-13-6-2-1-5-12(13)14(17)8-7-11-4-3-9-15-10-11/h1-10,16H/b8-7+

InChI key

CRWNZUBUBIULHB-BQYQJAHWSA-N

Biochem/physiol Actions

NSC49652 is a orally active, potent and selective inhibitor of death receptor p75NTR (p75 neurotrophin receptor) that targets the interfaces between transmembrane domains (TMDs). NSC49652 induces profound conformational changes and triggers p75NTR dependent cancer cell death. It inhibits the growth of cancer in melanoma mouse model.
orally active, potent and selective inhibitor of death receptor p75NTR that targets the interfaces between transmembrane domains

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eddy T H Goh et al.
Cell chemical biology, 25(12), 1485-1494 (2018-10-09)
Small molecules offer powerful ways to alter protein function. However, most proteins in the human proteome lack small-molecule probes, including the large class of non-catalytic transmembrane receptors, such as death receptors. We hypothesized that small molecules targeting the interfaces between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service