Skip to Content
Merck
All Photos(1)

Key Documents

SML2506

Sigma-Aldrich

Regadenoson

≥98% (HPLC)

Synonym(s):

2-[4-[(Methylamino)carbonyl]-1H-pyrazol-1-yl]adenosine, CVT-3146

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€110.00

€110.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€110.00

About This Item

Empirical Formula (Hill Notation):
C15H18N8O5
CAS Number:
Molecular Weight:
390.35
UNSPSC Code:
12352200
NACRES:
NA.77

€110.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]4(ncc(c4)C(=O)NC)c1nc2[n](cnc2c(n1)N)[C@@H]3O[C@@H]([C@H]([C@H]3O)O)CO

InChI

1S/C15H18N8O5/c1-17-13(27)6-2-19-23(3-6)15-20-11(16)8-12(21-15)22(5-18-8)14-10(26)9(25)7(4-24)28-14/h2-3,5,7,9-10,14,24-26H,4H2,1H3,(H,17,27)(H2,16,20,21)/t7-,9-,10-,14-/m1/s1

InChI key

LZPZPHGJDAGEJZ-AKAIJSEGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A2A-Adenosine receptor agonist; coronary vasodilator
Regadenoson (CVT-3146) is an A2A adenosine receptor agonist that is a coronary vasodilator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service