Skip to Content
Merck
All Photos(1)

Key Documents

SML2497

Sigma-Aldrich

BIO 1211 trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

2-MPUPA-LDVP trifluoroacetate, 4-(N′-(2-Methylphenyl)urea)-phenylacetyl-LDVP trifluoroacetate, BIO-1211 trifluoroacetate, BIO1211 trifluoroacetate, N-[2-[4-[[[(2-Methylphenyl)amino]carbonyl]amino]phenyl]acetyl]-L-leucyl-L-α-aspartyl-L-valyl-L-proline trifluoroacetate, N-[[4-[[[(2-Methylphenyl)amino]carbonyl]amino]-phenyl]acetyl]-LDVP trifluoroacetate, N-[[4-[[[(2-Methylphenyl)amino]carbonyl]amino]-phenyl]acetyl]-Leu-Asp-Val-Pro trifluoroacetate, N-[[4-[[[(2-Methylphenyl)amino]carbonyl]amino]-phenyl]acetyl]-fibronectin CS-1 fragment (1980-1983) trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H48N6O9 · xC2HF3O2
CAS Number:
Molecular Weight:
708.80 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

film
lyophilized powder

storage condition

desiccated

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

N3([C@@H](CCC3)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)Cc1ccc(cc1)NC(=O)Nc2c(cccc2)C)CC(C)C)CC(=O)O)C(C)C

InChI

1S/C36H48N6O9/c1-20(2)17-26(38-29(43)18-23-12-14-24(15-13-23)37-36(51)40-25-10-7-6-9-22(25)5)32(46)39-27(19-30(44)45)33(47)41-31(21(3)4)34(48)42-16-8-11-28(42)35(49)50/h6-7,9-10,12-15,20-21,26-28,31H,8,11,16-19H2,1-5H3,(H,38,43)(H,39,46)(H,41,47)(H,44,45)(H,49,50)(H2,37,40,51)/t26-,27-,28-,31-/m0/s1

InChI key

NVVGCQABIHSJSQ-KFZSMJGVSA-N

Biochem/physiol Actions

BIO 1211 is a selective, tight-binding α4β1 (VLA-4, very late antigen-4; koff = 1.4 x 10-4/s, KD = 70 pM) integrin antagonist (VCAM-Ig Jurkat surface binding IC50 = 1 nM; integrin-mediated cell adhesion IC50 = 4 nM/α4β1, 2 μM/α4β7, >100 μM/α1β1, α5β1, α6β1, αLβ2, αIIBβ3) based on the Leu-Asp-Val (LDV) sequence from the alternatively spliced connecting segment-1 (CS-1) of fibronectin (aa 1980-1983). In addition to probing α4β1-mediated cellular responses, BIO 1211 is also widely used in animal disease models in vivo, including MS (5-10 mg/kg; murine EAE) and asthma (1-10 mg/kg via intranasal or nebulizer to mice, rats, sheep; 3 mg/sheep iv.).
Potent and selective α4β1 integrin (VLA-4) antagonist for probing α4β1-mediated cellular & pathological responses in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Abraham et al.
American journal of respiratory and critical care medicine, 162(2 Pt 1), 603-611 (2000-08-10)
The leukocyte integrin very late antigen-4 (alpha(4)beta(1), CD49d/CD29) is an adhesion receptor that plays an important role in allergic inflammation and contributes to antigen-induced late responses (LAR) and airway hyperresponsiveness (AHR). In this study, we show that single doses of
Olli-Pekka Pulkka et al.
Journal of cellular and molecular medicine, 22(4), 2220-2230 (2018-01-30)
The molecular mechanisms for the dissemination and metastasis of gastrointestinal stromal tumours (GIST) are incompletely understood. The purpose of the study was to investigate the clinical relevance of integrin alpha 4 (ITGA4) expression in GIST. GIST transcriptomes were first compared
Gloria C Koo et al.
American journal of respiratory and critical care medicine, 167(10), 1400-1409 (2003-02-06)
A nonpeptidyl small molecule antagonist, compound A, to nonactivated very late antigen-4 (VLA4) was examined in lung inflammation induced by a single dose of ovalbumin challenge. Compound A presented a good pharmacokinetic property, when given intratracheally, and the blood cells
B V Karanam et al.
Xenobiotica; the fate of foreign compounds in biological systems, 37(5), 487-502 (2007-05-25)
BIO1211 is a small peptidyl potent antagonist of the activated form of alpha4beta1 integrin. The effect of enalapril on the in vitro and in vivo cleavage of BIO1211 was investigated. In heparinized blood, plasma and rat liver, lung and intestinal
Wei-Sheng Chen et al.
Nature communications, 7, 11302-11302 (2016-04-14)
Lymphangiogenesis plays a pivotal role in diverse pathological conditions. Here, we demonstrate that a carbohydrate-binding protein, galectin-8, promotes pathological lymphangiogenesis. Galectin-8 is markedly upregulated in inflamed human and mouse corneas, and galectin-8 inhibitors reduce inflammatory lymphangiogenesis. In the mouse model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service