Skip to Content
Merck
All Photos(1)

Key Documents

SML2395

Sigma-Aldrich

Elocalcitol

≥98% (HPLC)

Synonym(s):

(1R,3Z,5S)-3-[(2E)-2-[(3aS,7aS)-1-[(1S,3E)-5-ethyl-5-hydroxy-1-methyl-3-hepten-1-yl]-3,3a,5,6,7,7a-hexahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-5-fluoro-4-methylenecyclohexanol, BXL 628, BXL-628, Ro 26-9228

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€105.00
5 MG
€310.00

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€105.00
5 MG
€310.00

About This Item

Empirical Formula (Hill Notation):
C29H43FO2
CAS Number:
Molecular Weight:
442.65
MDL number:
UNSPSC Code:
12352200

€105.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +97 to +107°, c = c=1 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

F[C@H]1C[C@@H](C\C(=C\C=C2\[C@H]3[C@](CCC\2)(C(=CC3)[C@H](C\C=C\C(O)(CC)CC)C)C)\C1=C)O

InChI

1S/C29H43FO2/c1-6-29(32,7-2)17-8-10-20(3)25-14-15-26-22(11-9-16-28(25,26)5)12-13-23-18-24(31)19-27(30)21(23)4/h8,12-14,17,20,24,26-27,31-32H,4,6-7,9-11,15-16,18-19H2,1-3,5H3/b17-8+,22-12+,23-13-/t20-,24+,26-,27-,28+/m0/s1

InChI key

LRLWXBHFPGSUOX-GJQYOBCGSA-N

Biochem/physiol Actions

Elocalcitol (BX-L628) is an analog of Vitamin D3 that acts as an agonist at vitamin D VDR receptors. It has been shown to inhibit prostate cell growth and RhoA/Rho-kinase signaling. Elocalcitol (BXL-628) downregulated the expression of active RhoA and ROCK activity induced by high glucose and was shown to have renal protective effects.
Elocalcitol is a Vitamin D receptor (VDR) agonist and is an analog of calcitriol.[1][2] Elocalcitol possesses anti-inflammatory and anti- proliferative properties.[1] It regulates cell apoptosis and cell proliferation by binding to Vitamin D receptors (VDR).[3] It shows a therapeutic effect on people suffering from Benign prostatic hyperplasia (BPH) and lower urinary tract symptoms (LUTS).[2]
Vitamin D3 analog

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elocalcitol, a vitamin D3 analog for the potential treatment of benign prostatic hyperplasia, overactive bladder and male infertility.
Tiwari A.
IDrugs : The Investigational Drugs Journal, 12(6), 381-393 (2012)
Cristina Antinozzi et al.
The Journal of steroid biochemistry and molecular biology, 167, 169-181 (2017-01-04)
Vitamin D plays a pivotal role to maintain skeletal muscle integrity and health. Vitamin D deficiency characterizes inflammatory myopathy (IM) and diabetes, often overlapping diseases involving skeletal muscle damage. Vitamin D receptor (VDR) agonists likely exert beneficial effects in both
Wei Zhang et al.
Experimental and therapeutic medicine, 13(5), 1969-1976 (2017-06-02)
It has previously been reported that 1,25-(OH)2D3 inhibits high glucose-induced epithelial-to-mesenchymal transition (EMT) in HK-2 cells. However, the mechanism of this renoprotective action remains unclear. Elocalcitol (BXL-628), a vitamin D analog, has been suggested to be effective on the RhoA/Rho
The selective vitamin D receptor agonist, elocalcitol, reduces endometriosis development in a mouse model by inhibiting peritoneal inflammation
Mariani M, et al.
Human Reproduction, 27(7), 2010-2019 (2012)
Elocalcitol (BXL-628): a novel, investigational therapy for the therapeutic management of benign prostatic hyperplasia
Tiwari A.
Expert Opinion on Investigational Drugs, 17(5), 819-824 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service