Skip to Content
Merck
All Photos(1)

Key Documents

SML2206

Sigma-Aldrich

SKA-121

≥98% (HPLC)

Synonym(s):

5-Methylnaphth[2,1-d]oxazol-2-amine, 5-Methylnaphtho[2,1-d]oxazol-2-amine, SKA 121, SKA121

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.50
25 MG
€320.00

€99.50


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
€99.50
25 MG
€320.00

About This Item

Empirical Formula (Hill Notation):
C12H10N2O
CAS Number:
Molecular Weight:
198.22
UNSPSC Code:
12352200
NACRES:
NA.77

€99.50


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

form

powder

color

brown to dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C1=C2C=CC=C1)=CC3=C2OC(N)=N3

InChI

1S/C12H10N2O/c1-7-6-10-11(15-12(13)14-10)9-5-3-2-4-8(7)9/h2-6H,1H3,(H2,13,14)

InChI key

JEGUERWMMMQFJV-UHFFFAOYSA-N

Biochem/physiol Actions

KCa3.1 subtype-selective Ca2+-activated K+ channel activator with in vivo arterial blood pressure-lowering efficacy. More potent and selective than SKA-31.
SKA-121 is a KCa3.1 (IKCa1, IK1, KCa4, SKCa4, SK4) subtype-selective small conductance Ca2+-activated K+ channel (SK channel) positive-gating modulator (EC50 = 109 nM/KCa3.1 vs. 8.7 μM/KCa2.1, 6.8 μM/KCa2.2, 4.4 μM/KCa2.3) with 200- to 400-fold selectivity over CaV1.2 as well as representative KVs (KV1.3, KV2.1, KV3.1, and KV11.1) and NaV (NaV1.2, NaV1.4, NaV1.5, and NaV1.7) channels. When applied in vivo, SKA-121 significantly lowers mean arterial blood pressure in wild-type, but not KCa3.1(-/-), normotensive and hypertensive mice (100 mg/kg i.p.). SKA-121 is a more potent and selective KCa3.1 activator than SKA-31 (EC50 = 260 nM/KCa3.1, 2.9 μM/KCa2.1, 1.9 μM/KCa2.2, 2.9 μM/KCa2.3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aida Oliván-Viguera et al.
Basic & clinical pharmacology & toxicology, 119(2), 184-192 (2016-01-29)
Opening of intermediate-conductance calcium-activated potassium channels (KC a 3.1) produces membrane hyperpolarization in the vascular endothelium. Here, we studied the ability of two new KC a 3.1-selective positive-gating modulators, SKA-111 and SKA-121, to (1) evoke porcine endothelial cell KC a
Aida Oliván-Viguera et al.
Frontiers in physiology, 8, 39-39 (2017-02-16)
The calcium/calmodulin-gated KCa3.1 channel regulates normal and abnormal mitogenesis by controlling K
Palle Christophersen et al.
Channels (Austin, Tex.), 9(6), 336-343 (2015-07-29)
This short review discusses pharmacological modulation of the opening/closing properties (gating) of small- and intermediate-conductance Ca(2+)-activated K(+) channels (KCa2 and KCa3.1) with special focus on mechanisms-of-action, selectivity, binding sites, and therapeutic potentials. Despite KCa channel gating-modulation being a relatively novel
Brandon M Brown et al.
Molecular pharmacology, 92(4), 469-480 (2017-08-02)
Intermediate-conductance (K
Nichole Coleman et al.
Molecular pharmacology, 86(3), 342-357 (2014-06-25)
Small-conductance (KCa2) and intermediate-conductance (KCa3.1) calcium-activated K(+) channels are voltage-independent and share a common calcium/calmodulin-mediated gating mechanism. Existing positive gating modulators like EBIO, NS309, or SKA-31 activate both KCa2 and KCa3.1 channels with similar potency or, as in the case

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service