Skip to Content
Merck
All Photos(1)

Key Documents

SML2024

Sigma-Aldrich

GSK4028

≥97% (HPLC)

Synonym(s):

(S,S)-32, 4-Bromo-2-methyl-5-(((3S,5S)-1-methyl-5-phenylpiperidin-3-yl)amino)pyridazin-3(2H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€113.00
25 MG
€450.00

€113.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€113.00
25 MG
€450.00

About This Item

Empirical Formula (Hill Notation):
C17H21BrN4O
CAS Number:
Molecular Weight:
377.28
UNSPSC Code:
12352200
NACRES:
NA.77

€113.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(C[C@@H](C1)C2=CC=CC=C2)C[C@H]1NC(C=NN(C3=O)C)=C3Br

InChI key

VZAFGXCWAWRULT-KGLIPLIRSA-N

Biochem/physiol Actions

GSK4028 is an enantiomeric negative control for PCAF/GCN5 bromodomain inhibitor GSK4027.
enantiomeric negative control for GSK4027

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service