Skip to Content
Merck
All Photos(1)

Key Documents

SML1970

Sigma-Aldrich

PKUMDL-WQ-2101

≥98% (HPLC)

Synonym(s):

2,4-Dihydroxy-benzoic acid 2-[(2-hydroxy-5-nitrophenyl)methylene]hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€120.00
25 MG
€481.00

€120.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€120.00
25 MG
€481.00

About This Item

Empirical Formula (Hill Notation):
C14H11N3O6
CAS Number:
Molecular Weight:
317.25
UNSPSC Code:
12352200
NACRES:
NA.77

€120.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PKUMDL-WQ-2101 is a potent and selective non-NAD+-competing allosteric inhibitor of phosphoglycerate dehydrogenase (PHGDH) that selectively inhibits serine synthesis in cancer cells. PKUMDL-WQ-2101 binds to allosteric site I, which is located close to active site. PKUMDL-WQ-2101 inhibits tumor growth of MDA-MB-468 xenografts in mice.
Potent and selective non-NAD+-competing allosteric inhibitor of phosphoglycerate dehydrogenase (PHGDH)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service