Skip to Content
Merck
All Photos(1)

Documents

SML1926

Sigma-Aldrich

GAT211

≥98% (HPLC)

Synonym(s):

3-(2-Nitro-1-phenylethyl)-2-phenyl-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N2O2
CAS Number:
Molecular Weight:
342.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=[N+]([O-])CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3)NC4=CC=CC=C42

Biochem/physiol Actions

GAT211 is a positive allosteric modulator (PAM) of cannabinoid CB1 receptor signaling. GAT211 was found to amplify the therapeutic effect of endocannabinoids without the negative side effects of psychoactivity or tolerance. GAT211 caused a CB1-dependent suppression of paclitaxel-induced mechanical and cold allodynia with no sign of tolerance over 19 days of administration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service