Skip to Content
Merck
All Photos(1)

Key Documents

SML1783

Sigma-Aldrich

BAY-364

≥98% (HPLC)

Synonym(s):

6-(3-Hydroxypropyl)-2-(1-methyl-2-oxo-2,3-dihydro-1H-benzimidazol-5-yl)-1H-benzo[de]isoquinoline-1,3(2H)-dione, BAY-299N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19N3O4
CAS Number:
Molecular Weight:
401.41
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=C2C(C(CCCO)=CC=C23)=CC=C1)N(C4=CC=C(N(C)C(N5)=O)C5=C4)C3=O

Biochem/physiol Actions

BAY-364 (BAY-299N) is structurally similar to selective BRD1/TAF1 inhibitor BAY-299 and serves as inactive control.
BAY-364 (BAY-299N) is structurally similar to selective BRD1/TAF1 inhibitor BAY-299 and serves as inactive control. BAY-364 is inactive against BRD1 and exhibit moderate activity against TAF1 (3 μM). For full characterization details of the active probe, please visit the BAY-299 probe summary on the Structural Genomics Consortium (SGC) website.

BAY-299, the active probe, is available from Sigma. To learn more about and purchase BAY-299, click here.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc

Features and Benefits

BAY-364 is a negative control for BAY-299, an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service