Skip to Content
Merck
All Photos(1)

Key Documents

SML1591

Sigma-Aldrich

Buclizine dihydrochloride

≥98% (HPLC)

Synonym(s):

1-(p-Chloro-α-phenylbenzyl)-4-(p-tert-butylbenzyl)piperazine, 1-(p-Chlorobenzhydryl)-4-(p-tert-butylbenzyl)-diethylenediamine hydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]-4-[[4-(1,1-dimethylethyl)phenyl]methyl]-piperazine dihydrochloride, Buclina, Longifene, Posdel, UCB-4445, Vibazine, Vibazine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€114.00

€114.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€114.00

About This Item

Empirical Formula (Hill Notation):
C28H33ClN2 · 2HCl
CAS Number:
Molecular Weight:
505.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€114.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Clc1ccc(cc1)C(N3CCN(CC3)Cc4ccc(cc4)C(C)(C)C)c2ccccc2.[H+].[H+]

InChI

1S/C28H33ClN2.2ClH/c1-28(2,3)25-13-9-22(10-14-25)21-30-17-19-31(20-18-30)27(23-7-5-4-6-8-23)24-11-15-26(29)16-12-24;;/h4-16,27H,17-21H2,1-3H3;2*1H

InChI key

SDBHDSZKNVDKNU-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Biochem/physiol Actions

Buclizine is a piperazine antihistamine and anticholinergic that exerts its antiemetic and antimotion sickness effects.
Histamine H1 receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service