Skip to Content
Merck
All Photos(1)

Key Documents

SML1437

Sigma-Aldrich

Pimecrolimus

≥97% (HPLC)

Synonym(s):

33-epi-chloro-33-desoxyascomycin, Picrolimus

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€123.00
50 MG
€493.00

€123.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€123.00
50 MG
€493.00

About This Item

Empirical Formula (Hill Notation):
C43H68ClNO11
CAS Number:
Molecular Weight:
810.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€123.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@H]([C@@H](O)CC([C@H](CC)/C=C1C)=O)[C@](/C(C)=C/[C@@]2([H])C[C@@H](OC)[C@@H](Cl)CC2)([H])OC([C@@]3([H])N(C(C([C@@]4(O)[C@H](C)C[C@H](OC)[C@@H]([C@@H](OC)C[C@@H](C)C\1)O4)=O)=O)CCCC3)=O

InChI

1S/C43H68ClNO11/c1-10-30-18-24(2)17-25(3)19-36(53-8)39-37(54-9)21-27(5)43(51,56-39)40(48)41(49)45-16-12-11-13-32(45)42(50)55-38(28(6)33(46)23-34(30)47)26(4)20-29-14-15-31(44)35(22-29)52-7/h18,20,25,27-33,35-39,46,51H,10-17,19,21-23H2,1-9H3/b24-18+,26-20+/t25-,27+,28+,29-,30+,31-,32-,33-,35+,36-,37-,38+,39+,43+/m0/s1

InChI key

KASDHRXLYQOAKZ-XDSKOBMDSA-N

Gene Information

human ... FKBP1A(2280)

Biochem/physiol Actions

Calcineurin inhibitor, immune suppressant
Pimecrolimus is known for its anti-inflammatory and immunomodulatory properties and its low systemic immunosuppressive potential.[1]
Pimecrolimus, also known as Picrolimus, is a macrolactam ascomycin derivative that acts as an immune suppressant. It binds with high affinity to macrophilin-12 (FKBP-12) and inhibits calcineurin, a calcium-dependent phosphatase. Pimecrolimus inhibits production of pro-inflammatory cytokines by T cells and mast cells and prevents the release of inflammatory cytokines and mediators from mast cells. It has been used clinically as a treatment for eczema.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Gupta et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 17(5), 493-503 (2003-08-28)
Pimecrolimus (SDZ ASM 981), an ascomycin derivative, is one of the new classes of immunomodulating macrolactams and was specifically developed for the treatment of inflammatory skin diseases. The interest in pimecrolimus has been substantial because of its significant anti-inflammatory activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service