Skip to Content
Merck
All Photos(1)

Key Documents

SML1397

Sigma-Aldrich

β-Naltrexamine dihydrochloride

≥98% (HPLC)

Synonym(s):

(5α,6β)-6-Amino-17-(cyclopropylmethyl)-4,5α-epoxy-morphinan-3,14-diol dihydrochloride, 6β-Naltrexamine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€649.00

€649.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€649.00

About This Item

Empirical Formula (Hill Notation):
C20H26N2O3 · 2HCl
CAS Number:
Molecular Weight:
415.35
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

€649.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@]1(CC[C@@H](N)[C@]2([H])OC3=C4O)[C@]52C3=C(C=C4)C[C@H]1N(CC6CC6)CC5

InChI

1S/C20H26N2O3/c21-13-5-6-20(24)15-9-12-3-4-14(23)17-16(12)19(20,18(13)25-17)7-8-22(15)10-11-1-2-11/h3-4,11,13,15,18,23-24H,1-2,5-10,21H2/t13-,15-,18+,19+,20-/m1/s1

InChI key

SPPAUICKSNQRNC-GNUVVZJLSA-N

Biochem/physiol Actions

β-Naltrexamine is a potent opioid antagonist.
β-Naltrexamine is a potent orally available, long lasting opioid antagonist.
β-naltrexamine is obtained from naltrexone.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morgan Le Naour et al.
Journal of medicinal chemistry, 55(2), 670-677 (2011-12-06)
Using the selective mu-kappa agonist, N-naphthoyl-β-naltrexamine 1, as the prototype ligand, a series of closely related naphthalene analogues were synthesized to study the chemical space around the naphthalene moiety in an effort to evaluate how receptor selectivity is affected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service