Skip to Content
Merck
All Photos(1)

Key Documents

SML1336

Sigma-Aldrich

VUAA1

≥98% (HPLC)

Synonym(s):

2-((4-Ethyl-5-(pyridin-3-yl)-4H-1,2,4-triazol-3-yl)thio)-N-(4-ethylphenyl)acetamide, N-(4-Ethylphenyl)-2-((4-ethyl-5-(3-pyridinyl)-4H-1,2,4-triazol-3-yl)thio)acetamide, VU0099414

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€438.00

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€438.00

About This Item

Empirical Formula (Hill Notation):
C19H21N5OS
CAS Number:
Molecular Weight:
367.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCC(C=C1)=CC=C1NC(CSC2=NN=C(C3=CN=CC=C3)N2CC)=O

InChI

1S/C19H21N5OS/c1-3-14-7-9-16(10-8-14)21-17(25)13-26-19-23-22-18(24(19)4-2)15-6-5-11-20-12-15/h5-12H,3-4,13H2,1-2H3,(H,21,25)

InChI key

UWCCKVJVOHTHAF-UHFFFAOYSA-N

Biochem/physiol Actions

N-(4-ethylphenyl)-2-((4-ethyl-5-(3-pyridinyl)-4H-1,2,4-triazol-3-yl) thio) acetamide (VUAA1) is a non-naturally occurring[1] volatile compound.[2] It is considered as a valuable tool for insect olfactory research.[1]
VUAA1 is an agonist of the highly conserved insect odorant receptor co-receptor ion channel Orco.
VUAA1 is an agonist of the highly conserved insect odorant receptor co-receptor ion channel Orco. VUAA1 is able to activate both heteromeric and homomeric Orco-containing channels.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endogenous insensitivity to the Orco agonist VUAA1 reveals novel olfactory receptor complex properties in the specialist fly Mayetiola destructor.
Corcoran JA, et al.
Scientific Reports, 8(1), 3489-3489 (2018)
Expressional and functional interactions of two Apis cerana cerana olfactory receptors.
Guo L, et al.
PeerJ, 6, e5005-e5005 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service