Skip to Content
Merck
All Photos(1)

Documents

SML1320

Sigma-Aldrich

NSC687852

≥98% (HPLC)

Synonym(s):

(3E,5E)-1-Acryloyl-3,5-bis(4-nitrobenzylidene)piperidin-4-one, 3,5-Bis((4-nitrophenyl)methylidene)-1-prop-2-enoylpiperidin-4-one, b-AP15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3O6
CAS Number:
Molecular Weight:
419.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(C(CN(C(C=C)=O)C1)=CC2=CC=C([N+]([O-])=O)C=C2)C1=CC3=CC=C([N+]([O-])=O)C=C3

InChI

1S/C22H17N3O6/c1-2-21(26)23-13-17(11-15-3-7-19(8-4-15)24(28)29)22(27)18(14-23)12-16-5-9-20(10-6-16)25(30)31/h2-12H,1,13-14H2

InChI key

GFARQYQBWJLZMW-UHFFFAOYSA-N

Biochem/physiol Actions

NSC687852 is a cell-permeable, potent, reversible and selective inhibitor of 19S regulatory-particle-associated deubiquitinases (DUBs) UCHL5 and USP14, with no effect on UCH-L1, UCH-L3, USP2, USP7, USP8 and BAP1 and on the proteasomal proteolytic activities. NSC687852 induces cell-cycle arrest and apoptosis in several colon cancers and in CNS cells, and suppresses tumor growth in various mouse models.

Other Notes

Light and air sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service