Skip to Content
Merck
All Photos(1)

Documents

SML1312

Sigma-Aldrich

Mubritinib

≥98% (HPLC)

Synonym(s):

(E)-4-((4-(4-(1H-1,2,3-triazol-1-yl)butyl)phenoxy)methyl)-2-(4-(trifluoromethyl)styryl)oxazole, 1-[4-[4-[[2-[(1E)-2-[4-(Trifluoromethyl)phenyl]ethenyl]-4-oxazolyl]methoxy]phenyl]butyl]-1H-1,2,3-triazole, CID 6444692, TAK 165, TAK-165

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23F3N4O2
CAS Number:
Molecular Weight:
468.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)C1=CC=C(/C=C/C2=NC(COC3=CC=C(C=C3)CCCCN4C=CN=N4)=CO2)C=C1

InChI

1S/C25H23F3N4O2/c26-25(27,28)21-9-4-20(5-10-21)8-13-24-30-22(18-34-24)17-33-23-11-6-19(7-12-23)3-1-2-15-32-16-14-29-31-32/h4-14,16,18H,1-3,15,17H2/b13-8+

InChI key

ZTFBIUXIQYRUNT-MDWZMJQESA-N

Gene Information

human ... ERBB2(2064)

Biochem/physiol Actions

Mubritinib (TAK-165) is a potent and selective inhibitor of the human epidermal growth factor receptor 2 ErbB (HER2) tyrosine kinase. Mubritinib potently inhibits proliferation of cancer cell lines over- expressing ErbB2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxana V Denisova et al.
Scientific reports, 12(1), 13796-13796 (2022-08-14)
Therapeutic resistance to kinase inhibitors constitutes a major unresolved clinical challenge in cancer and especially in glioblastoma. Multi-kinase inhibitors may be used for simultaneous targeting of multiple target kinases and thereby potentially overcome kinase inhibitor resistance. However, in most cases
Ravi Holani et al.
Gut microbes, 12(1), 1785802-1785802 (2020-07-14)
We hypothesized that the antimicrobial peptide cathelicidin has a physiological role in regulating gut inflammatory homeostasis. We determined that cathelicidin synergizes with LPS to facilitate its internalization and signaling via endosomic TLR4 in colonic epithelium, evoking synthesis of the human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service