Skip to Content
Merck
All Photos(1)

Key Documents

SML1223

Sigma-Aldrich

XBD173

≥98% (HPLC)

Synonym(s):

AC-5216, Emapunil, N-Ethyl-7,8-dihydro-7-methyl-8-oxo-2-phenyl-N-(phenylmethyl)-9H-purine-9-acetamide, NVP-XBD173

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€438.00

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€438.00

About This Item

Empirical Formula (Hill Notation):
C23H23N5O2
CAS Number:
Molecular Weight:
401.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(C1=CN=C(C2=CC=CC=C2)N=C1N3CC(N(CC)CC4=CC=CC=C4)=O)C3=O

InChI

1S/C23H23N5O2/c1-3-27(15-17-10-6-4-7-11-17)20(29)16-28-22-19(26(2)23(28)30)14-24-21(25-22)18-12-8-5-9-13-18/h4-14H,3,15-16H2,1-2H3

InChI key

NBMBIEOUVBHEBM-UHFFFAOYSA-N

Biochem/physiol Actions

XBD173 (emapunil, AC-5216) is a potent and selective translocator protein 18 kDa (TSPO) ligand with a Ki value of 0.297 nM. XBD173 acts at the TSPO translocator protein, formerly called the peripheral benzodiazepine receptor, which is involved in production of neuroactive steroids such as allopregnanolone in the brain. TSPO carries cholesterol from the outer to the inner mitochondrial membrane, which is the rate-limiting step of steroidogenesis. XBD173 enhances GABA-mediated neurotransmission via induction of neurosteroidogenesis and exhibits anti-anxiety and antidepressant-like effects.
XBD173 is a potent and selective translocator protein.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service