Skip to Content
Merck
All Photos(1)

Key Documents

SML1186

Sigma-Aldrich

GGTI-286 dihydrochloride

≥90% (HPLC)

Synonym(s):

(R)-N-[[5-[(2-Amino-3-mercaptopropyl)amino][1,1′-biphenyl]-2-yl]carbonyl]-L-lLeucine methyl ester dihydrochloride, GGTI 286 dihydrochloride, N-[[5-[[(2R)-2-Amino-3-mercaptopropyl]amino][1,1′-biphenyl]-2-yl]carbonyl]-L-leucine methyl ester dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€228.00
25 MG
€578.00

€228.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€228.00
25 MG
€578.00

About This Item

Empirical Formula (Hill Notation):
C23H31N3O3S · 2HCl
CAS Number:
Molecular Weight:
502.50
UNSPSC Code:
12352200
NACRES:
NA.77

€228.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥90% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

SC[C@H](N)CNc1cc(c(cc1)C(=O)N[C@@H](CC(C)C)C(=O)OC)c2ccccc2

InChI key

IENQPUVVSDIXCT-UTKZUKDTSA-N

Biochem/physiol Actions

GGTI-286 is a cell permeable, potent and selective inhibitor of geranylgeranyltransferase I (GGTase I) that selectively disrupts oncogenic K-Ras4B. New study shows that GGTI-286 reduces nuclear localization of β-catenin.
GGTI-286 is a cell permeable, potent and selective inhibitor of geranylgeranyltransferase I (GGTase I). CAAX peptidomimetic.

Other Notes

Air sensitive; dessicant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service