Skip to Content
Merck
All Photos(1)

Key Documents

SML1065

Sigma-Aldrich

TR100

≥98% (HPLC)

Synonym(s):

2-Cyano-3-[1-[3-(dimethylamino)propyl]-2-methyl-1H-indol-3-yl]-N-octyl-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€157.00
25 MG
€402.00

€157.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€157.00
25 MG
€402.00

About This Item

Empirical Formula (Hill Notation):
C26H38N4O
CAS Number:
Molecular Weight:
422.61
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€157.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=C(/C=C(C#N)\C(NCCCCCCCC)=O)C2=C(N1CCCN(C)C)C=CC=C2

InChI

1S/C26H38N4O/c1-5-6-7-8-9-12-16-28-26(31)22(20-27)19-24-21(2)30(18-13-17-29(3)4)25-15-11-10-14-23(24)25/h10-11,14-15,19H,5-9,12-13,16-18H2,1-4H3,(H,28,31)/b22-19-

InChI key

XXRLIGWROLDOLS-QOCHGBHMSA-N

Biochem/physiol Actions

TR100 is a specific anti-tropomyosin agent that preferentially disrupts the actin cytoskeleton of tumor cells.
TR100 is a specific anti-tropomyosin agent that preferentially disrupts the actin cytoskeleton of tumor cells. TR100 selectively disrupts Tm5NM1/2 containing actin filaments. TR100 induces apoptosis in a panel of neural crest-derived tumor cell lines with no adverse effects on cardiac structure and function.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Baron et al.
PloS one, 11(11), e0164284-e0164284 (2016-11-11)
Quantitative magnetic resonance imaging (qMRI) offers several advantages in imaging and determination of soft tissue alterations when compared to qualitative imaging techniques. Although applications in brain and muscle tissues are well studied, its suitability to quantify relaxation times of intact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service