Skip to Content
Merck
All Photos(1)

Key Documents

SML1059

Sigma-Aldrich

MMB-4

≥98% (HPLC)

Synonym(s):

1,1′-Methylenebis[4-[(hydroxyimino)methyl]-pyridinium dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€114.00
25 MG
€451.00

€114.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€114.00
25 MG
€451.00

About This Item

Empirical Formula (Hill Notation):
C13H14N4O2 · 2Br
CAS Number:
Molecular Weight:
418.08
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€114.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

light yellow to light brown

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O/N=C/C1=CC=[N+](C[N+]2=CC=C(/C=N\O)C=C2)C=C1.[Br-].[Br-]

InChI

1S/C13H12N4O2.2BrH/c18-14-9-12-1-5-16(6-2-12)11-17-7-3-13(4-8-17)10-15-19;;/h1-10H,11H2;2*1H

InChI key

ZKQNRRLCBJUEBC-UHFFFAOYSA-N

Biochem/physiol Actions

MMB-4 is also called as 1,1′-Methylenebis{4-[(hydroxyimino)methyl]pyridinium) dichloride. It serves as a promising antidote for organophosphate poisoning.[1]
MMB-4 is an oxime that can reactivate cholinesterases that have been inactivated by exposure to organophosphates such as sarin or soman. Administration of MMB-4 and atropine normalizes many respiratory parameters in guinea pigs following soman inhalation.
MMB-4 is an oxime that can reactivate cholinesterases.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Macauley et al.
Journal of pharmaceutical and biomedical analysis, 49(4), 889-894 (2009-02-17)
1,1'-Methylenebis{4-[(hydroxyimino)methyl]pyridinium) dichloride (MMB-4), a promising antidote for organophosphate poisoning, has been shown by chromatography and NMR to be a mixture of geometric isomers, predominantly the E/E form. The chromatographically separated isomers have been isolated, directly characterized by NMR to be

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service