Skip to Content
Merck
All Photos(1)

Key Documents

SML0899

Sigma-Aldrich

Windorphen

≥98% (HPLC)

Synonym(s):

(Z)-3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO3
CAS Number:
Molecular Weight:
302.75
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C17H15ClO3/c1-20-14-7-3-12(4-8-14)16(11-19)17(18)13-5-9-15(21-2)10-6-13/h3-11H,1-2H3/b17-16-

InChI key

VNRALGZMXHFBPG-MSUUIHNZSA-N

Biochem/physiol Actions

Windorphen is a selective Wnt/β-catenin signaling inhibitor acting through selective inhibition of p300, a histone acetyltransferase (HAT) that also acts as a critical coactivator of β-catenin, a transcription factor that mediates the Wnt signaling pathway involved in embryogenesis and cancer cell survival. Windorphen blocks Wnt signaling by selectively disrupting the association of p300 with the C-terminal transactivation domain of β-catenin-1. Windorphen does not inhibit the closely related CREB-binding protein (CBP). Windorphen induced apoptosis in several Wnt-dependent tumor cell lines including colon adenocarcinoma SW480and RKO and prostate cancer cell lines DU145 and PC3.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service