Skip to Content
Merck
All Photos(1)

Key Documents

SML0755

Sigma-Aldrich

LDN-192960 dihydrochloride

≥98% (HPLC)

Synonym(s):

3-[(2,7-Dimethoxy-9-acridinyl)thio]-1-propanamine dihydrochloride, 3-[(2,7-Dimethoxyacridin-9-yl)thio]propan-1-amine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00

€127.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€127.00

About This Item

Empirical Formula (Hill Notation):
C18H20N2O2S · 2HCl
CAS Number:
Molecular Weight:
401.35
UNSPSC Code:
12352200
NACRES:
NA.77

€127.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

S(CCCN)c1c2c(nc3c1cc(cc3)OC)ccc(c2)OC

InChI

1S/C18H20N2O2S/c1-21-12-4-6-16-14(10-12)18(23-9-3-8-19)15-11-13(22-2)5-7-17(15)20-16/h4-7,10-11H,3,8-9,19H2,1-2H3

InChI key

ZFOMCSNUEHMROO-UHFFFAOYSA-N

Biochem/physiol Actions

LDN-192960 is a potent and selective inhibitor of haspin (Haploid Germ Cell-Specific Nuclear Protein Kinase).
LDN-192960 is a potent and selective inhibitor of haspin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service