Skip to Content
Merck
All Photos(1)

Key Documents

SML0672

Sigma-Aldrich

VU0405601

≥98% (HPLC)

Synonym(s):

2-[(1-Bromo-2-naphthalenyl)oxy]-N-3-pyridinyl-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€297.00

€297.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€297.00

About This Item

Empirical Formula (Hill Notation):
C17H13BrN2O2
CAS Number:
Molecular Weight:
357.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€297.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=C2C=CC=CC2=CC=C1OCC(NC3=CC=CN=C3)=O

InChI

1S/C17H13BrN2O2/c18-17-14-6-2-1-4-12(14)7-8-15(17)22-11-16(21)20-13-5-3-9-19-10-13/h1-10H,11H2,(H,20,21)

InChI key

WLLRAGBPCSOJNH-UHFFFAOYSA-N

Biochem/physiol Actions

VU0405601 is a hERG (the human Ether-à-go-go-Related Gene) agonist that increased the IC50 of dofetilide from 38.7 nM to 76.3 nM. VU0405601 protects against arrhythmias induced by hERG inhibition.
VU0405601 is a hERG agonist that protects against arrhythmias induced by hERG inhibition.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service