Skip to Content
Merck
All Photos(1)

Key Documents

SML0588

Sigma-Aldrich

CP-94,253 dihydrochloride

≥98% (HPLC)

Synonym(s):

1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride, 3- (1,2,5,6-Tetrahydro-4-pyridyl)-5-propoxypyrrolo[3, 2-b] pyridine hydrochloride, 5-Propoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-pyrrolo[3,2-b]pyridine hydrochloride, CP-94253 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.10
25 MG
€275.00

€96.10


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
€96.10
25 MG
€275.00

About This Item

Empirical Formula (Hill Notation):
C15H19N3O · 2HCl
CAS Number:
Molecular Weight:
330.25
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€96.10


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.Cl.CCCOc1ccc2[nH]cc(C3=CCNCC3)c2n1

InChI

1S/C15H19N3O.2ClH/c1-2-9-19-14-4-3-13-15(18-14)12(10-17-13)11-5-7-16-8-6-11;;/h3-5,10,16-17H,2,6-9H2,1H3;2*1H

InChI key

HGFPLQPNCZQRLO-UHFFFAOYSA-N

Biochem/physiol Actions

CP-94,253 is a potent and selective serotonin 5-HT1B receptor agonist with approximately 25x and 40x selectivity over the closely related 5-HT1D and 5-HT1A receptors. Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively. CP-94,253 was found to be anxiolytic in animal models; dose-dependently enhanced reinforcing effects of cocaine (self-administration);and reduced food intake.
CP-94,253 is a potent and selective serotonin 5-HT1B receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

  1. Is this product blood-brain barrier permeable?

    1 answer
    1. The compound is blood-brain barrier permeable, as indicated by a review published in J Physiol. 2009 Jan 1; 587(Pt 1): 49–60, titled "Pharmacological targeting of the serotonergic system for the treatment of obesity" (PMID: 19029184)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service