Skip to Content
Merck
All Photos(1)

Key Documents

SML0453

Sigma-Aldrich

TBPB

≥98% (HPLC)

Synonym(s):

1-(1′-(2-Methylbenzyl)-1,4′-bipiperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.60

€96.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€96.60

About This Item

Empirical Formula (Hill Notation):
C25H32N4O
CAS Number:
Molecular Weight:
404.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€96.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution; warmed)

storage temp.

2-8°C

SMILES string

Cc1ccccc1CN2CCC(CC2)N3CCC(CC3)N4C(=O)Nc5ccccc45

InChI

1S/C25H32N4O/c1-19-6-2-3-7-20(19)18-27-14-10-21(11-15-27)28-16-12-22(13-17-28)29-24-9-5-4-8-23(24)26-25(29)30/h2-9,21-22H,10-18H2,1H3,(H,26,30)

InChI key

CWPKTBMRVATCBL-UHFFFAOYSA-N

Biochem/physiol Actions

TBPB is a highly selective Muscarinic M1 receptor allosteric agonist.
TBPB is a highly selective Muscarinic M1 receptor allosteric agonist. It activates M1 through an allosteric site rather than the orthosteric acetylcholine binding site, which is likely critical for its selectivity. Muscarinic 1 (M1) receptors are expressed in brain regions responsible for attention and memory, including hippocampus, cortex, and striatum. M1 activation is a proposed mechanism for increasing information processing in disease states, such as Alzheimer′s. M1 agonists are being studied as potential therapeutic agents to treat Alzheimer′s disease and the cognitive and negative symptoms of schizophrenia.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwini Kumar et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 115(1), 3-10 (2015-07-30)
Alzheimer's disease (AD) is a neurodegenerative disorder in which the death of brain cells causes memory loss and cognitive decline, i.e., dementia. The disease starts with mild symptoms and gradually becomes severe. AD is one of the leading causes of
Hidenori Hashimoto et al.
Scientific reports, 7(1), 17216-17216 (2017-12-10)
Ionic clathrate hydrates can selectively capture small gas molecules such as CO
Aleksei Furletov et al.
Sensors (Basel, Switzerland), 20(17) (2020-09-02)
Nowadays, analytical systems based on silver triangular nanoplates (AgTNPs) have been shown as good prospects for chemical sensing. However, they still remain relatively poorly studied as colorimetric probes for sensing various classes of compounds. This study shows that these nanoparticles
Shang-Chi Lin et al.
Scientific reports, 4, 6976-6976 (2014-11-14)
A good diagnostic procedure avoids wasting medical resources, is easy to use, resists contamination, and provides accurate information quickly to allow for rapid follow-up therapies. We developed a novel diagnostic procedure using a "cotton-based diagnostic device" capable of real-time detection
Leonardo M Dos Santos et al.
Journal of environmental management, 268, 110340-110340 (2020-05-10)
CO2 separation from natural gas is considered to be a crucial strategy to mitigate global warming problems, meet product specification, pipeline specs and other application specific requirements. Silica xerogels (SX) are considered to be potential materials for CO2 capture due

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service