Skip to Content
Merck
All Photos(1)

Key Documents

SML0343

Sigma-Aldrich

Methylstat

≥98% (HPLC)

Synonym(s):

(2E)-4-[Hydroxy[4-[[[4-[[[(1-naphthalenylamino)carbonyl]oxy]methyl]phenyl]methyl]amino]butyl]amino]-4-oxo-2-butenoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€196.00
25 MG
€774.00

€196.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€196.00
25 MG
€774.00

About This Item

Empirical Formula (Hill Notation):
C28H31N3O6
CAS Number:
Molecular Weight:
505.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€196.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC2=C1C=CC=C2)OCC3=CC=C(CNCCCCN(O)C(/C=C/C(OC)=O)=O)C=C3

InChI

1S/C28H31N3O6/c1-36-27(33)16-15-26(32)31(35)18-5-4-17-29-19-21-11-13-22(14-12-21)20-37-28(34)30-25-10-6-8-23-7-2-3-9-24(23)25/h2-3,6-16,29,35H,4-5,17-20H2,1H3,(H,30,34)/b16-15+

InChI key

MUJOCHRZXRZONW-FOCLMDBBSA-N

Biochem/physiol Actions

Methylstat is an inhibitor of the Jumonji C domain-containing histone trimethyl demethylases (JHDMs).
Methylstat is an inhibitor of the Jumonji C domain-containing histone trimethyl demethylases (JHDMs). Methylstat inhibits the activity of JMJD2C (IC50 = 4.3 μM), with similar potency against JMJD2E and JMJD3. The compound has poor activity against dimethyl demethylases, and does not inhibit class I or II HDACs. Methylsat blocks proliferation of the esophageal carcinoma cell line KYSE150 with an IC50 value of 5.1 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Fellous et al.
Genes, 10(9) (2019-09-13)
Histone methylation patterns are important epigenetic regulators of mammalian development, notably through stem cell identity maintenance by chromatin remodeling and transcriptional control of pluripotency genes. But, the implications of histone marks are poorly understood in distant groups outside vertebrates and
Qingsong Hu et al.
Nature immunology, 20(7), 835-851 (2019-06-05)
How tumor cells genetically lose antigenicity and evade immune checkpoints remains largely elusive. We report that tissue-specific expression of the human long noncoding RNA LINK-A in mouse mammary glands initiates metastatic mammary gland tumors, which phenotypically resemble human triple-negative breast
Qingsong Hu et al.
Cell research, 29(4), 286-304 (2019-01-12)
Despite the structural conservation of PTEN with dual-specificity phosphatases, there have been no reports regarding the regulatory mechanisms that underlie this potential dual-phosphatase activity. Here, we report that K27-linked polyubiquitination of PTEN at lysines 66 and 80 switches its phosphoinositide/protein
Evdokiia Potolitsyna et al.
Communications biology, 7(1), 458-458 (2024-04-16)
Differentiation of adipose progenitor cells into mature adipocytes entails a dramatic reorganization of the cellular architecture to accommodate lipid storage into cytoplasmic lipid droplets. Lipid droplets occupy most of the adipocyte volume, compressing the nucleus beneath the plasma membrane. How
Emily O Wisniewski et al.
Science advances, 6(31), eaba6505-eaba6505 (2020-08-14)
How migrating cells differentially adapt and respond to extracellular track geometries remains unknown. Using intravital imaging, we demonstrate that invading cells exhibit dorsoventral (top-to-bottom) polarity in vivo. To investigate the impact of dorsoventral polarity on cell locomotion through different confining

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service